missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Estrogen Receptor beta Mouse anti-Human, Clone: MC10, eBioscience™

Mouse Monoclonal Antibody

$120.12 - $308.88

Specifications

Antigen Estrogen Receptor beta
Clone MC10
Host Species Murine
Species Reactivity Human
Applications ELISA,Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunoprecipitation,Microscopy,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122778 Life Technologies
LS14933680
25μg Each for $120.12
501122779 Life Technologies
LS14933682
100μg Each for $308.88
Specifications

Description

The MC10 monoclonal antibody recognizes all forms of human estrogen receptor (ER) beta. ER beta is a member of a superfamily of nuclear receptors, which are ligand-dependent transcription factors. ER beta like ER alpha, becomes activated by binding the hormone estrogen, causing a dissociation from chaperone proteins and subsequent dimerization with either ER beta or ER alpha. Unlike ER alpha, the result of ER beta signaling is thought to be anti-proliferative and proapoptotic. The MC10 clone recognizes full length ER beta as well as each of its splice variants.
Description & Specifications

Specifications

Estrogen Receptor beta
Murine
ELISA,Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunoprecipitation,Microscopy,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
MC10
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer