Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-21, Rat Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and ELISA experiments with Thermo Scientific™ Estrogen Receptor Ab-21, Rat Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RT1641P0

 View more versions of this product

Catalog No. RT1641P0

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Format Concentrated
Immunogen Estrogen receptor purified from MCF-7 cells
Cross Reactivity Human
Clone H222
Quantity 100µL
Regulatory Status RUO
Antigen Estrogen Receptor Ab-21
Host Species Rat
Isotype IgG1 kappa
Conjugate Unlabeled
Research Discipline Oncogenesis
Type Primary
Classification Monoclonal
Concentration 200µg/mL

Ensure accurate, reproducible results in western blotting and ELISA experiments with Thermo Scientific™ Estrogen Receptor Ab-21, Rat Monoclonal Antibody.

ER protein exhibits six functionally discrete domains required for transcription activation, binding to estrogen response element (ERE), constitutive dimerization, binding to heat shock proteins, and ligand recognition. It is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors may improve response to anti-estrogen therapy.

Host Species: Rat

Clone: H222

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Shows broad species reactivity.

Immunogen: Estrogen receptor purified from MCF-7 cells

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • ELISA

Note:

USA: RUO; Int'l: RUO