missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

$272.73 - $1,371.63

Specifications

Antigen Estrogen Receptor Ab-17
Concentration 1mg/mL
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1521P Thermo Scientific™ Lab Vision™
RB1521P
Purified Ab with BSA and Azide 1mL Each for $1,248.73
RB1521P0 Thermo Scientific™ Lab Vision™
RB1521P0
Purified Ab with BSA and Azide 100μL Each for $272.73
RB1521P1 Thermo Scientific™ Lab Vision™
RB1521P1
Purified Ab with BSA and Azide 500μL Each for $755.69
RB1521P1ABX Thermo Scientific™ Lab Vision™
RB1521P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $848.22
RB1521PABX Thermo Scientific™ Lab Vision™
RB1521PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,371.63
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Estrogen Receptor Ab-17
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Unlabeled
Rabbit
Steroids Hormones and Growth Factors
A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein
1mg/mL
Polyclonal
Concentrated
RUO
Primary
IgG