Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  RB1521PABX

 View more versions of this product

Catalog No. RB1521PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Quantity 1mL
Format Concentrated
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG
Antigen Estrogen Receptor Ab-17
Regulatory Status RUO
Immunogen A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein
Host Species Rabbit
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Research Discipline Steroids Hormones and Growth Factors
Classification Polyclonal
Concentration 1mg/mL
Item Description 1mL; 1mg/mL; Unlabeled; Purified without BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody.

ER gene consists of more than 140kb of genomic DNA divided into 8 exons, being translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO