missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer:  Thermo Scientific™ Lab Vision™ RB1521P1ABX

 View more versions of this product

Catalog No. RB1521P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Estrogen Receptor Ab-17, Rabbit Polyclonal Antibody.

ER gene consists of more than 140kb of genomic DNA divided into 8 exons, being translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse, Rabbit, Sheep, Pig and Cow. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Estrogen Receptor Ab-17
Polyclonal
Unlabeled
Concentrated
A synthetic peptide from the N-terminus of human estrogen receptor, alpha protein
500μL
Steroids Hormones and Growth Factors
Polyclonal
Immunoprecipitation,Western Blotting
1mg/mL
Ab without BSA/Azide (Frozen)
Rabbit
IgG
RUO
Primary
Bovine,Human,Murine,Porcine,Rabbit,Rat