Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-16, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

$268.82 - $1,357.16

Specifications

Antigen Estrogen Receptor Ab-16
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1493P

 
thermo scientific™ lab vision™
RB1493P
Purified Ab with BSA and Azide 1mL Each for $1,258.96

RB1493P0

 
thermo scientific™ lab vision™
RB1493P0
Purified Ab with BSA and Azide 100µL Each for $268.82

RB1493P1

 
thermo scientific™ lab vision™
RB1493P1
Purified Ab with BSA and Azide 500µL Each for $753.80

RB1493P1ABX

 
thermo scientific™ lab vision™
RB1493P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $832.02

RB1493PABX

 
thermo scientific™ lab vision™
RB1493PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,357.16

RB1493R7

 
thermo scientific™ lab vision™
RB1493R7
Ready-to-use for IHC (UVLP) 7mL Each for $721.06
Description & Specifications

Specifications

Antigen Estrogen Receptor Ab-16
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Hamster
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen A synthetic peptide derived from the C-terminus of human estrogen receptor, alpha protein
Isotype IgG

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Estrogen Receptor Ab-16, Rabbit Polyclonal Antibody.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Hamster. Does not react with rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human estrogen receptor, α protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Gel Supershift (Not verified)

Note:

USA: RUO; Int'l: RUO