Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Estrogen Receptor Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in affinity purification, gel supershift and immunoprecipitation experiments.

$244.65 - $1,317.27

Specifications

Antigen Estrogen Receptor Ab-1
Clone AER314
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Bovine
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS168B

 
thermo scientific™ lab vision™
MS168B
200μg/mL Biotin 1mL Each for $1,317.27

MS168B0

 
thermo scientific™ lab vision™
MS168B0
200μg/mL Biotin 100µL Each for $340.22

MS168B1

 
thermo scientific™ lab vision™
MS168B1
200μg/mL Biotin 500µL Each for $894.71

MS168P

 
thermo scientific™ lab vision™
MS168P
200μg/mL Unlabeled 1mL Each for $1,028.86

MS168P0

 
thermo scientific™ lab vision™
MS168P0
200μg/mL Unlabeled 100µL Each for $244.65

MS168P1

 
thermo scientific™ lab vision™
MS168P1
200μg/mL Unlabeled 500µL Each for $679.57

MS168P1ABX

 
thermo scientific™ lab vision™
MS168P1ABX
1mg/mL Unlabeled 100µL Each for $770.06

MS168PABX

 
thermo scientific™ lab vision™
MS168PABX
1mg/mL Unlabeled 200µL Each for $1,088.37
Description & Specifications

Specifications

Antigen Estrogen Receptor Ab-1
Clone AER314
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Purified, SDS-denatured calf uterus estrogen receptor (ER)
Isotype IgG1 lambda

Ensure accurate, reproducible results in affinity purification, gel supershift and immunoprecipitation experiments with Thermo Scientific™ Estrogen Receptor Ab-1, Mouse Monoclonal Antibody.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: AER314

Isotype: IgG1/λ

Species Reactivity: Human, Cow, Rabbit, Mouse and Rat. Others not known.

Epitope: aa 125-165

Immunogen: Purified, SDS-denatured calf uterus estrogen receptor (ER)

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Affinity Purification
  • Gel Supershift
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO