Please login to your online account to display your discounted pricing

Estriol, >97%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0215107691

 View more versions of this product

Catalog No. ICN15107691

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material Estriol
CAS 50-27-1
Physical Form White crystalline powder
Assay Percent Range >97%
Molecular Formula C18H24O3
Formula Weight 288.4g/mol
Synonym 3, 16α, 17β-Trihydroxy-1

Specifications

Chemical Name or Material Estriol
CAS 50-27-1
Physical Form White crystalline powder
Assay Percent Range >97%
Molecular Formula C18H24O3
Formula Weight 288.4g/mol
Synonym 3, 16α, 17β-Trihydroxy-1