Please login to your online account to display your discounted pricing

Estriol, >97%, MP Biomedicals™


Quantity 100mg

Manufacturer: mp biomedicals  15107680

 View more versions of this product

Catalog No. ICN15107680

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 288.4
Color White
Quantity 100mg
Assay Percent Range >97%
Chemical Name or Material Estriol
Molecular Formula C18H24O3
CAS 50-27-1
Synonym 3, 16α, 17β-Trihydroxy-1

Specifications

Chemical Name or Material Estriol
CAS 50-27-1
Physical Form Crystalline Powder
Assay Percent Range >97%
Molecular Formula C18H24O3
Formula Weight 288.4
Synonym 3, 16α, 17β-Trihydroxy-1