Please login to your online account to display your discounted pricing

Estriol, >97%, MP Biomedicals™

$40.16 - $40.16

Chemical Identifiers

Chemical Name or Material Estriol
CAS 50-27-1
Physical Form Crystalline Powder
Assay Percent Range >97%
Molecular Formula C18H24O3
Formula Weight 288.4
Synonym 3, 16α, 17β-Trihydroxy-1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15107680

 
mp biomedicals
15107680
100mg Each for $40.16
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 288.4
Color White
Quantity 100mg
Assay Percent Range >97%
Chemical Name or Material Estriol
Molecular Formula C18H24O3
CAS 50-27-1
Synonym 3, 16α, 17β-Trihydroxy-1