Please login to your online account to display your discounted pricing

Esterase Bs3 From Bacillus Stearothermophilus, Acros Organics


Manufacturer: thermo fisher scientific  AC381011000

 View more versions of this product

Catalog No. AC381011000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Name Esterase BS3
Quantity 100mg
Storage Requirements 4°C
Source Bacillus stearothermophilus
Format Solid
Concentration >2.5U/mg
Color Light Gray