Please login to your online account to display your discounted pricing

Esterase Bs3 From Bacillus Stearothermophilus, Acros Organics

Specifications

Color Light Gray
Concentration >2.5U/mg
Product Name Esterase BS3
Format Solid
Source Bacillus stearothermophilus
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC381010250

 
fisher scientific acros
AC381010250
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381011000

 
thermo fisher scientific
AC381011000
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Light Gray
Concentration >2.5U/mg
Product Name Esterase BS3
Format Solid
Source Bacillus stearothermophilus
Storage Requirements 4°C