Please login to your online account to display your discounted pricing

ESAM Rat anti-Mouse, PE, Clone: 1G8, eBioscience™

Rat Monoclonal Antibody

$172.55 - $274.05

Specifications

Antigen ESAM
Clone 1G8
Host Species Rat
Gene Alias Endothelial cell-Selective Adhesion Molecule
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010904

 
life technologies
LS12585281
50μg Each for $172.55

5010905

 
life technologies
LS12585282
100μg Each for $274.05
Description & Specifications

Specifications

Antigen ESAM
Clone 1G8
Host Species Rat
Gene Alias Endothelial cell-Selective Adhesion Molecule
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 1G8 monoclonal antibody reacts with mouse ESAM (endothelial cell-selective adhesion molecule). ESAM is a 55kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.