Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythrosin Yellowish

$85.58 - $299.85

Chemical Identifiers

Chemical Name or Material Erythrosin Yellowish
CAS 33239-19-9
Molecular Formula C20H8I2Na2O5
Formula Weight 628.07
MDL Number MFCD00036461
Synonym C.I. 45425; Acid red 95
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1562614

  SDS
alfa aesar™
A1562614
25g Each for $85.58

AAA1562622

  SDS
alfa aesar™
A1562622
100g Each for $299.85
Description & Specifications

Specifications

Formula Weight 628.07
Chemical Name or Material Erythrosin Yellowish
Quantity 25g
Molecular Formula C20H8I2Na2O5
MDL Number MFCD00036461
CAS 33239-19-9
Synonym C.I. 45425; Acid red 95
Melting Point >300°C