Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythropoietin (EPO)(117-131), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67594ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67594ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Erythropoietin (EPO) (117-131), Certified Mass Spec Grade
Format Lyophilized powder
Storage Requirements -30° to -10°C
Sequence EAISPPDAASAAPLR
Molecular Weight 1465.60
Purification Method HPLC
Quantity 3 vials, 500 pmol/vial
Regulatory Status RUO