Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythromycin Resistance Peptide

For cellular and molecular biology applications

Specifications

Product Type Erythromycin Resistance Peptide
Molecular Weight 664.86
Storage Requirements -30° to -10°C
Sequence Met-Arg-Leu-Phe-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66230MA

 
alfa aesar™
J66230MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66230MB

 
alfa aesar™
J66230MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Erythromycin Resistance Peptide
Molecular Weight 664.86
Storage Requirements -30° to -10°C
Sequence Met-Arg-Leu-Phe-Val
Format Solid
Common Name MRLFV
Regulatory Status RUO
Purification Method HPLC