Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythromycin ethylsuccinate


Quantity 25g

Manufacturer: alfa aesar™  J6654314

 View more versions of this product

Catalog No. AAJ6654314

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 862.05
Melting Point 219° to 224°C
Assay 95%
Health Hazard 1 Danger
CAS 1264-62-6
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
MDL Number MFCD00084692
RTECS Number WM9800000
Solubility Information Freely soluble in organic solvents, practically insoluble in water
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Color White
Quantity 25g
Chemical Name or Material Erythromycin ethylsuccinate

Specifications

Chemical Name or Material Erythromycin ethylsuccinate
CAS 1264-62-6
Formula Weight 862.05
MDL Number MFCD00084692

  • A broad spectrum antibiotic commonly targeting bacteria responsible for respiratory tract infections

Accessories

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

SDS & Safety Documents