Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythromycin ethylsuccinate

Chemical Identifiers

Chemical Name or Material Erythromycin ethylsuccinate
CAS 1264-62-6
Formula Weight 862.05
MDL Number MFCD00084692
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6654314

  SDS
alfa aesar™
J6654314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 862.05
Melting Point 219° to 224°C
Assay 95%
CAS 1264-62-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Physical Form Powder
MDL Number MFCD00084692
Solubility Information Freely soluble in organic solvents, practically insoluble in water
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Quantity 25g
RTECS Number WM9800000
Chemical Name or Material Erythromycin ethylsuccinate

  • A broad spectrum antibiotic commonly targeting bacteria responsible for respiratory tract infections

Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.