Please login to your online account to display your discounted pricing

Alfa Aesar™ Erythromycin estolate, 95%

A macrolide antibiotic with a target spectrum similar to penicillin

Chemical Identifiers

Chemical Name or Material Erythromycin estolate
CAS 3521-62-8
Formula Weight 1056.38
MDL Number MFCD00084691
Merck Index 14,3683
Synonym Erythromycin 2'-propionate dodecyl sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6654806

 
alfa aesar™
J6654806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 135° to 140°C
Formula Weight 1056.38
Chemical Name or Material Erythromycin estolate
Assay 95%
CAS 3521-62-8
RTECS Number KF5775000
Synonym Erythromycin 2'-propionate dodecyl sulfate
Physical Form Powder
Merck Index 14,3683
MDL Number MFCD00084691
Solubility Information Freely soluble in organic solvents, practically insoluble in water
Quantity 5g
Color White to off-white
Boiling Point 743°C
Sensitivity Light Sensitive