Please login to your online account to display your discounted pricing

Erythroid Cells Mouse anti-Rat, FITC, Clone: HIS49, eBioscience™

Mouse Monoclonal Antibody

$361.05 - $361.05

Specifications

Antigen Erythroid Cells
Clone HIS49
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122185

 
life technologies
LS11975582
100μg Each for $361.05
Description & Specifications

Specifications

Antigen Erythroid Cells
Clone HIS49
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody HIS49 reacts with an unknown antigen expressed by rat erythroid cells and their progenitors. HIS49 does not react with lymphocytes, monocytes or granulocytes.