Please login to your online account to display your discounted pricing

(±)-erythro-Aleuritic acid, 95%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  133731000

 View more versions of this product

Catalog No. AC133731000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 304.41
Quantity 100g
Color White to beige
EINECS Number 208-578-8
TSCA TSCA
RTECS Number ML9850000
Beilstein 03, III, 901
Merck Index 15, 225
CAS Min % 94.0
CAS Max % 100.0
Synonym 9, 10, 16-Trihydroxypalmitic acid
Chemical Name or Material (±)-erythro-Aleuritic acid
Assay Percent Range 95%
Molecular Formula C16H32O5
CAS 533-87-9
Name Note 95%
Linear Formula HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
MDL Number MFCD00135596
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (±)-erythro-Aleuritic acid
Name Note 95%
CAS 533-87-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C16H32O5
Linear Formula HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
Formula Weight 304.41
MDL Number MFCD00135596
Synonym 9, 10, 16-Trihydroxypalmitic acid