Please login to your online account to display your discounted pricing

(±)-erythro-Aleuritic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (±)-erythro-Aleuritic acid
Name Note 95%
CAS 533-87-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C16H32O5
Linear Formula HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
Formula Weight 304.41
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC133730250

  SDS
thermo fisher scientific
133730250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC133731000

  SDS
thermo fisher scientific
133731000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 304.41
Quantity 25g
Color White to beige
Beilstein 03, III, 901
Merck Index 15, 225
EINECS Number 208-578-8
TSCA TSCA
RTECS Number ML9850000
CAS Min % 94.0
CAS Max % 100.0
Synonym 9, 10, 16-Trihydroxypalmitic acid
Chemical Name or Material (±)-erythro-Aleuritic acid
Linear Formula HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
MDL Number MFCD00135596
CAS 533-87-9
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C16H32O5
Packaging Glass bottle