missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ertapenem sodium salt, 100mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific J67047XF

 View more versions of this product

Catalog No. AAJ67047XF



Description

Description

Applications
Ertapenem has in vitro activity against gram-positive and gram-negative aerobic and anaerobic bacteria. The bactericidal activity of ertapenem results from the inhibition of cell wall synthesis and is mediated through ertapenem binding to penicillin binding proteins.

Solubility
Fully miscible in water. Soluble in DMSO.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Specifications

Ertapenem sodium salt
C22H24N3NaO7S
Plastic bottle
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
131675376
Liquid
153773-82-1
MFCD09475566
Fully miscible in water. Soluble in DMSO.
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
497.498
CHEBI:60070
1mL

Chemical Identifiers

153773-82-1
497.498
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
CHEBI:60070
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
C22H24N3NaO7S
MFCD09475566
131675376
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

Documents