Please login to your online account to display your discounted pricing

Eriochrome™ Blue-Black R, Reagent Grade, LabChem™

$52.77 - $52.77

Chemical Identifiers

Chemical Name or Material Eriochrome™ Blue-Black R
CAS 2538-85-4
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula HOC10H6N:NC10H5OHSO3H
Formula Weight 416.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC140558

 
lab chem
LC140558
Amber Glass 25g Each for $52.77
Description & Specifications

Specifications

Formula Weight 416.39
Chemical Name or Material Eriochrome™ Blue-Black R
Quantity 25g
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula HOC10H6N:NC10H5OHSO3H
CAS 2538-85-4
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Black
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature