Please login to your online account to display your discounted pricing

Eriochrome™ Blue-Black R, Dry Form in NaCl, for Calcium, Certified, LabChem™

$41.77 - $41.77

Chemical Identifiers

Chemical Name or Material Eriochrome™ Blue-Black R
CAS 7647-14-5
CAS Max % 99.8
CAS 2538-85-4
CAS Max % 0.2
Name Note Dry Form in NaCl, for Calcium
Purity Grade Certified Grade
Linear Formula HOC10H6N:NC10H5OHSO3H
Formula Weight 416.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC140609

 
lab chem
LC140609
Poly Bottle 100g Each for $41.77
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 416.39
Chemical Name or Material Eriochrome™ Blue-Black R
Quantity 100g
EINECS Number 231-598-3
Name Note Dry Form in NaCl, for Calcium
Physical Form Grains
Linear Formula HOC10H6N:NC10H5OHSO3H
CAS 7647-14-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.8
Health Hazard 2 GHS H Statement
Product is not hazardous.
CAS 2538-85-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 2.165g/cm3
Suitability Passes Test
CAS Max % 0.2
Recommended Storage Room Temperature
Decomposition Information Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides; Sulfur compounds
Density 2.165g/cm3
Boiling Point 1,413°C
Melting Point 801°C
Color Purple
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature