Please login to your online account to display your discounted pricing

Eriochrome™ Black T, ACS Grade, LabChem™

$82.69 - $82.69

Chemical Identifiers

Chemical Name or Material Eriochrome™ Black T
CAS 1787-61-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C20H12N3O7NaS
Formula Weight 461.38
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC140358

 
lab chem
LC140358
Amber Glass 25g Each for $82.69
Description & Specifications

Specifications

Formula Weight 461.38
Chemical Name or Material Eriochrome™ Black T
Quantity 25g
EINECS Number 217-250-3
Purity Grade ACS Grade
Physical Form Powder
Linear Formula C20H12N3O7NaS
CAS 1787-61-7
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Suitability Passes Test
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Black
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

Room Temperature