missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erie Garnet, ca. 80% dye content

$35.86 - $477.18

Chemical Identifiers

CAS 2429-70-1
Molecular Formula C32H21N5O7S2Na2
Formula Weight 697.65
MDL Number MFCD00046442
Synonym C.I. 22145; Congo Corinth
Chemical Name or Material Erie Garnet, ≈80% dye content
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1421606 Alfa Aesar™
A1421606
5g Each for $35.86
AAA1421614 Alfa Aesar™
A1421614
25g Each for $146.54
AAA1421622 Alfa Aesar™
A1421622
100g Each for $477.18
Specifications Description & Specifications

Chemical Identifiers

2429-70-1
697.65
C.I. 22145; Congo Corinth
C32H21N5O7S2Na2
MFCD00046442
Erie Garnet, ≈80% dye content

Specifications

2429-70-1
697.65
C.I. 22145; Congo Corinth
Erie Garnet, ≈80% dye content
C32H21N5O7S2Na2
MFCD00046442
5g

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning