Ergodyne™ Ergodyne™ Trex 6300 Ice Traction Device
Eight heat-treated carbon steel studs provide tenacious grip and stability
Manufacturer:  Ergodyne™ 16754
 View more versions of this product
Catalog No. 19-130-5945
Description
Description
  • Stretchable rubber for fit and easy on/off
  • Flexible to -40°C (-40°F)
  • Color: black
Specifications
Specifications
Ice traction device for shoes or boots
Trex 6300 ice traction device
Rubber
2 Replacement studs
Black
Large
Documents