Ergodyne™ Ergodyne™ GloWear 8385 Class III 4-in-1 Jackets
Manufacturer:  Ergodyne™ 24379
 View more versions of this product
Catalog No. 19-147-470
Specifications
Specifications
Storm Front, Zipper, Snap
ANSI ISEA 107 Class 3, Level 2
Flagging crew, road construction, utility crew, survey crew, construction, railway workers
Inset hood with drawstring and stoppers, removable sleeves
2 in. 3M™ Scotchlite™ reflective stripes
8.0
147 to 157cm
Polyurethane
Orange
5X-Large
300 Denier Oxford
Thinsulate™
58 to 62 in.
Documents