Please login to your online account to display your discounted pricing
Ergodyne™ Chill-Its™ 6200S Phase Change Cooling Vests With Packs
Comfortable vests maintain body temperature
$247.43 - $456.37
Specifications
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Color Size Price Quantity    
19-088-1747
ergodyne
12102
Clear Small/Medium Each for $247.43
19-088-1749
ergodyne
12112*
Clear Large/XL Each for $310.18
29-635-5401
ergodyne
12110
Khaki Large/XL Each for $456.37
Description & Specifications
Specifications
Quantity Each
  • Cooling lifecycle lasts up to four hours while maintaining a constant 14°C (58°F)
  • Banox FR3™ fabric provides an ATPV of 9.4cal/cm2
  • Nontoxic, noncarcinogenic, dielectric substance
  • Cotton construction with hook and loop closure
  • Pulp/Paper
  • Fabrication
  • Hazmat
  • Rescue
  • Vulcanizing
  • Construction
  • Refineries
  • Foundries