missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ergodyne™ Arsenal™ Gear Bags

Super-tough bags hold and separate turnout gear, equipment and personal items

Manufacturer:  Ergodyne™ 13305

 View more versions of this product

Catalog No. 19-130-3924




Description

Description

  • Built with heavy-duty #10 double-pull zippers
  • 1000D super-tough nylon repels water and resists abrasions, tears, scuffs and punctures
  • Capacity: 7280 cu. in.
  • Dimensions: 16 x 21 x 15 in.
  • 2 in. wide webbing handle and shoulder straps
  • Imprinted with Maltese Cross

Specifications

Specifications

1.8 lb.
Black
Gear Bag
0.81kg
Polyester, PVC
Each
Documents