Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ERCC1 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-2, Mouse Monoclonal Antibody.

$254.92 - $1,270.61

Specifications

Antigen ERCC1 Ab-2
Clone 8F1
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS671P

 
thermo scientific™ lab vision™
MS671P
200μg/mL 1mL Each for $1,192.47

MS671P0

 
thermo scientific™ lab vision™
MS671P0
200μg/mL 100μL Each for $254.92

MS671P1

 
thermo scientific™ lab vision™
MS671P1
200μg/mL 500μL Each for $716.49

MS671P1ABX

 
thermo scientific™ lab vision™
MS671P1ABX
1mg/mL 100μL Each for $779.59

MS671PABX

 
thermo scientific™ lab vision™
MS671PABX
1mg/mL 200μL Each for $1,270.61
Description & Specifications

Specifications

Antigen ERCC1 Ab-2
Clone 8F1
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant full length human ERCC1 protein
Isotype IgG2b

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-2, Mouse Monoclonal Antibody.

The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision. Elevated levels of ERCC1 have also been reported in Cisplatin-resistant cells.

Host Species: Mouse

Clone: 8F1

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human ERCC1 protein

Molecular Weight: 33-36 kDa

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO