Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ERCC1 Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS671P1ABX

 View more versions of this product

Catalog No. MS671P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Antigen ERCC1 Ab-2
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Clone 8F1
Immunogen Recombinant full length human ERCC1 protein
Isotype IgG2b
Conjugate Unlabeled
Research Discipline DNA Repair
Classification Monoclonal
Type Primary
Concentration 1mg/mL

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-2, Mouse Monoclonal Antibody.

The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision. Elevated levels of ERCC1 have also been reported in Cisplatin-resistant cells.

Host Species: Mouse

Clone: 8F1

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human ERCC1 protein

Molecular Weight: 33-36 kDa

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO