Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ERCC1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-1, Mouse Monoclonal Antibody.

$268.92 - $1,588.41

Specifications

Antigen ERCC1 Ab-1
Clone 3H11
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS647B

 
thermo scientific™ lab vision™
MS647B
200μg/mL Biotin 1mL Each for $1,588.41

MS647B0

 
thermo scientific™ lab vision™
MS647B0
200μg/mL Biotin 100µL Each for $364.29

MS647B1

 
thermo scientific™ lab vision™
MS647B1
200μg/mL Biotin 500µL Each for $991.04

MS647P

 
thermo scientific™ lab vision™
MS647P
200μg/mL Unlabeled 1mL Each for $1,259.18

MS647P0

 
thermo scientific™ lab vision™
MS647P0
200μg/mL Unlabeled 100µL Each for $268.92

MS647P1

 
thermo scientific™ lab vision™
MS647P1
200μg/mL Unlabeled 500µL Each for $761.65

MS647P1ABX

 
thermo scientific™ lab vision™
MS647P1ABX
1mg/mL Unlabeled 100µL Each for $846.39

MS647PABX

 
thermo scientific™ lab vision™
MS647PABX
1mg/mL Unlabeled 200µL Each for $1,383.49
Description & Specifications

Specifications

Antigen ERCC1 Ab-1
Clone 3H11
Applications Immunoprecipitation
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Full length recombinant human ERCC1 protein
Isotype IgG2a kappa

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-1, Mouse Monoclonal Antibody.

The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision. Elevated levels of ERCC1 have also been reported in Cisplatin-resistant cells.

Host Species: Mouse

Clone: 3H11

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Full-length recombinant human ERCC1 protein

Molecular Weight: 33-36 kDa

Positive Control: A431 or HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO