Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ ERCC1 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS647P1ABX

 View more versions of this product

Catalog No. MS647P1ABX

  • / Each
 

Description & Specifications

Specifications

Clone 3H11
Antigen ERCC1 Ab-1
Quantity 100µL
Regulatory Status RUO
Format Concentrated
Host Species Mouse
Isotype IgG2a kappa
Applications Immunoprecipitation
Immunogen Full length recombinant human ERCC1 protein
Cross Reactivity Human
Classification Monoclonal
Type Primary
Research Discipline DNA Repair
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ ERCC1 Ab-1, Mouse Monoclonal Antibody.

The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision. Elevated levels of ERCC1 have also been reported in Cisplatin-resistant cells.

Host Species: Mouse

Clone: 3H11

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Full-length recombinant human ERCC1 protein

Molecular Weight: 33-36 kDa

Positive Control: A431 or HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO