Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)

Chemical Identifiers

Chemical Name or Material Erbium rod
CAS 7440-52-0
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00010987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40288DH

  SDS
alfa aesar™
40288DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40288DJ

  SDS
alfa aesar™
40288DJ
50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40288HF

  SDS
alfa aesar™
40288HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cast Rod, dia. is approximate
Quantity 25mm
Packaging Mylar under Argon
Odor Odorless
Chemical Name or Material Erbium rod
Assay Percent Notes (metals basis excluding Ta)
MDL Number MFCD00010987
Storage Note 1
  • Packaged under argon
TSCA TSCA
Name Note 12.7mm (0.5 in.) dia.
Weight ≈1.2g/mm
Assay 99.9%
CAS 7440-52-0

  • Packaged under argon

Safety and Handling

  • Packaged under argon