missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium powder, -40 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4416906

 View more versions of this product

Catalog No. AA4416906



Description

Description

  • Packaged under Argon
Specifications

Specifications

Erbium powder
7440-52-0
Er
UN3089
chips
[Er]
167.259
CHEBI:33379
99.9%
Gray
5 g
(REO)
MFCD00010987
−40 Mesh
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
Powder
Air and moisture sensitive

Chemical Identifiers

7440-52-0
167.259
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
chips
CHEBI:33379
[Er]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-160-1

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Argon

Documents