missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium pieces, distilled dendritic, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1225503

 View more versions of this product

Catalog No. AA1225503



Description

Description

  • Packaged under argon
Specifications

Specifications

Erbium pieces
Silver
Distilled dendritic
(REO)
MFCD00010987
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
[Er]
167.259
CHEBI:33379
99.9%
Moisture Sensitive
1g
7440-52-0
Er
UN3178
UYAHIZSMUZPPFV-UHFFFAOYSA-N
erbium
23980
Pieces

Chemical Identifiers

7440-52-0
167.259
UYAHIZSMUZPPFV-UHFFFAOYSA-N
23980
erbium
Er
MFCD00010987
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
CHEBI:33379
[Er]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents