Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium pieces, distilled dendritic, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Erbium pieces
CAS 7440-52-0
Name Note Distilled dendritic
MDL Number MFCD00010987
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1225503

  SDS
alfa aesar™
1225503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1225506

  SDS
alfa aesar™
1225506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1225514

  SDS
alfa aesar™
1225514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Color Silver
CAS 7440-52-0
Sensitivity Moisture Sensitive
Name Note Distilled dendritic
Physical Form Pieces
Storage Note 1
  • Packaged under argon
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 1 Danger
TSCA TSCA
Chemical Name or Material Erbium pieces
Assay Percent Notes (REO)
UN Number UN3178
Assay 99.9%
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
MDL Number MFCD00010987
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

  • Packaged under argon