missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erbium Pieces, 99.9% (REO), Thermo Scientific Chemicals
$228.48 - $780.59
Chemical Identifiers
CAS | 7440-52-0 |
---|---|
Molecular Formula | Er |
Molecular Weight (g/mol) | 167.26 |
MDL Number | MFCD00010987 |
InChI Key | UYAHIZSMUZPPFV-UHFFFAOYSA-N |
Synonym | unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic |
PubChem CID | 23980 |
ChEBI | CHEBI:33379 |
IUPAC Name | erbium |
SMILES | [Er] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AA0011109
|
Thermo Scientific Chemicals
00011109 |
10 g | Glass Bottle under Argon |
Each for $228.48
|
|
||||
AA0011118
|
Thermo Scientific Chemicals
00011118 |
50 g | Packaged under argon in glass bottle |
Each for $780.59
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-52-0 | |
167.26 | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
23980 | |
erbium |
Er | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
CHEBI:33379 | |
[Er] |
Specifications
Silver to Gray | |
Silver Gray pieces | |
99.9% (REO) | |
MFCD00010987 | |
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic | |
[Er] | |
167.26 | |
CHEBI:33379 | |
Pieces |
7440-52-0 | |
9.066 g/cm3 | |
Er | |
10 g | |
UYAHIZSMUZPPFV-UHFFFAOYSA-N | |
erbium | |
23980 | |
Glass Bottle under Argon | |
Erbium, Rare Earth Oxides Content |
Safety and Handling
Keep container tightly closed and dry.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.1, Packaging Group: II
EINECSNumber : 231-160-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only