Please login to your online account to display your discounted pricing

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Erbium(III) perchlorate hexahydrate
CAS 14692-15-0
Purity Grade Reagent Grade
Molecular Formula Er(ClO4)3·6H2O
Formula Weight 573.70 (465.61 Anhydrous)
MDL Number MFCD00150232
UN Number UN3211
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA40576NC

  SDS
alfa aesar
40576NC
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 573.70 (465.61 Anhydrous)
Quantity g
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Purity Grade Reagent Grade
UN Number UN3211
Color Pink
Concentration 50% w/w in aq. soln.
Health Hazard 2 GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
Molecular Formula Er(ClO4)3·6H2O
MDL Number MFCD00150232
CAS 14692-15-0
Chemical Name or Material Erbium(III) perchlorate hexahydrate
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.