Learn More
Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040576NA
Description
Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Erbium(III) perchlorate hexahydrate | |
14692-15-0 | |
∼2 g | |
MFCD00150232 | |
Micible with water. | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] | |
573.687 | |
573.70 (465.61 Anhydrous) | |
Odorless |
Pink | |
Liquid | |
Cl3ErH12O18 | |
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade | |
JZRNHNTYRRYOCR-UHFFFAOYSA-K | |
erbium(3+);triperchlorate;hexahydrate | |
20038200 | |
Reagent | |
Pink liquid |
Chemical Identifiers
14692-15-0 | |
573.687 | |
JZRNHNTYRRYOCR-UHFFFAOYSA-K | |
20038200 | |
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] |
Cl3ErH12O18 | |
MFCD00150232 | |
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade | |
erbium(3+);triperchlorate;hexahydrate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
DOTInformation : DOT Class: 5.1, Packing Group: II
EINECSNumber : 237-840-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only