missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Erbium(III) perchlorate, 40 wt.% solution in water, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 428290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagentSpecifications
Erbium(III) perchlorate | |
5g | |
MFCD00016073 | |
FGBKFKCLWFRKEW-UHFFFAOYSA-K | |
erbium(3+);triperchlorate | |
15335705 | |
Glass bottle | |
1.4400g/mL |
14017-55-1,7732-18-5 | |
Cl3ErO12 | |
erbium 3+ perchlorate, erbium perchlorate, perchloric acid,erbium 3+ salt 8ci,9ci, erbium iii perchlorate solution, perchloric acid erbium iii salt, erbium perchlorate aqueous solution | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3] | |
465.61 | |
465.61 | |
1.44 |
RUO â Research Use Only