Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) oxide, REacton™, 99% (REO)

Chemical Identifiers

Chemical Name or Material Erbium(III) oxide
CAS 12061-16-4
Purity Grade Reacton™
Formula Weight 382.56
MDL Number MFCD00010991
Merck Index 14,3646
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA66222A1

  SDS
alfa aesar™
66222A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 382.56
Purity Grade Reacton™
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Sensitivity Hygroscopic
Melting Point 2400°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Merck Index 14,3646
Physical Form Powder
Assay 99%
CAS 12061-16-4
Health Hazard 1 Warning
MDL Number MFCD00010991
Chemical Name or Material Erbium(III) oxide
Odor Odorless
Assay Percent Notes (REO)
Density 8.64g/mL
Quantity 1kg
RTECS Number KD9250000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.