missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erbium(III) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

For vacuum deposition

Supplier:  Thermo Scientific Chemicals 03637222

 View more versions of this product

Catalog No. AA3637222


Description

Description

Erbium(III) oxide is used for vacuum deposition electrical, optical and photoluminescenc, gate dielectrics in semi conductor devices, and in Biomedicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Erbium(III) oxide
12061-16-4
100 g
Er2O3
14,3646
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
dierbium(3+) trioxidandiide
382.56
Hygroscopic
2400°C
Sintered Lump
(metals basis)
MFCD00010991
Soluble in water(0.0049g/L).
[O--].[O--].[O--].[Er+3].[Er+3]
382.52
99.8%
8.64 g/mL

Chemical Identifiers

12061-16-4
382.52
ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
[O--].[O--].[O--].[Er+3].[Er+3]
Er2O3
MFCD00010991
dierbium(3+) trioxidandiide
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 235-045-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only