missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) oxalate decahydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1321609

 View more versions of this product

Catalog No. AA1321609



Specifications

Specifications

Erbium(III) oxalate decahydrate
(REO)
Er2(C2O4)3·10H2O
UN3288
CYHKKMLSGVLNPD-UHFFFAOYSA-N
erbium;oxalic acid;decahydrate
131877482
Powder
Reacton™
Hygroscopic
30618-31-6
C6H26Er2O22
MFCD00150142
erbium oxalate decahydrate,
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Er].[Er]
784.77
778.73 (598.58 Anhydrous)
99.99%
2.64g/mL
10g

Chemical Identifiers

30618-31-6
784.77
CYHKKMLSGVLNPD-UHFFFAOYSA-N
131877482
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Er].[Er]
C6H26Er2O22
MFCD00150142
erbium oxalate decahydrate,
erbium;oxalic acid;decahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents