Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) oxalate decahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Erbium(III) oxalate decahydrate
CAS 30618-31-6
Purity Grade Reacton™
Linear Formula Er2(C2O4)3·10H2O
Formula Weight 778.73 (598.58 Anhydrous)
MDL Number MFCD00150142
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1321609

  SDS
alfa aesar™
1321609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1321618

  SDS
alfa aesar™
1321618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 778.73 (598.58 Anhydrous)
Chemical Name or Material Erbium(III) oxalate decahydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Powder
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula Er2(C2O4)3·10H2O
MDL Number MFCD00150142
CAS 30618-31-6
UN Number UN3288
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.64g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.