missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1130614

 View more versions of this product

Catalog No. AA1130614



Specifications

Specifications

Erbium(III) nitrate pentahydrate
(REO)
MFCD00149691
acmc-20al2v, er.3no3.5h2o, erbium iii nitrate pentahydrate, erbium iii nitrate pentahydrate, reacton, erbium nitrate pentahydrate, erbium nitrate, pentahydrate,, erbium nitrate-water 1/3/5, erbium trinitrate pentahydrate
LWHHUEHWVBVASY-UHFFFAOYSA-N
erbium(3+);trinitrate;pentahydrate
53249207
Crystalline Aggregates
Reacton™
Hygroscopic
25g
10031-51-3
ErH10N3O14
UN1477
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
443.346
443.35 (353.27 Anhydrous)
99.99%
Odorless
130°C (in -4H2O)

Chemical Identifiers

10031-51-3
443.346
LWHHUEHWVBVASY-UHFFFAOYSA-N
53249207
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
ErH10N3O14
MFCD00149691
acmc-20al2v, er.3no3.5h2o, erbium iii nitrate pentahydrate, erbium iii nitrate pentahydrate, reacton, erbium nitrate pentahydrate, erbium nitrate, pentahydrate,, erbium nitrate-water 1/3/5, erbium trinitrate pentahydrate
erbium(3+);trinitrate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 233-436-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents