missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

$224.91 - $658.30

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.346
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym acmc-20al2v, er.3no3.5h2o, erbium iii nitrate pentahydrate, erbium iii nitrate pentahydrate, reacton, erbium nitrate pentahydrate, erbium nitrate, pentahydrate,, erbium nitrate-water 1/3/5, erbium trinitrate pentahydrate
PubChem CID 53249207
IUPAC Name erbium(3+);trinitrate;pentahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1130614
View Documents
Alfa Aesar™
1130614
25g Each for $224.91
 
AA1130622
View Documents
Alfa Aesar™
1130622
100g Each for $658.30
 
Specifications

Chemical Identifiers

10031-51-3
443.346
LWHHUEHWVBVASY-UHFFFAOYSA-N
53249207
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
ErH10N3O14
MFCD00149691
acmc-20al2v, er.3no3.5h2o, erbium iii nitrate pentahydrate, erbium iii nitrate pentahydrate, reacton, erbium nitrate pentahydrate, erbium nitrate, pentahydrate,, erbium nitrate-water 1/3/5, erbium trinitrate pentahydrate
erbium(3+);trinitrate;pentahydrate

Specifications

10031-51-3
MFCD00149691
acmc-20al2v, er.3no3.5h2o, erbium iii nitrate pentahydrate, erbium iii nitrate pentahydrate, reacton, erbium nitrate pentahydrate, erbium nitrate, pentahydrate,, erbium nitrate-water 1/3/5, erbium trinitrate pentahydrate
LWHHUEHWVBVASY-UHFFFAOYSA-N
erbium(3+);trinitrate;pentahydrate
53249207
Crystalline Aggregates
Reacton™
Hygroscopic
25g
(REO)
ErH10N3O14
UN1477
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
443.346
443.35 (353.27 Anhydrous)
99.99%
Odorless
130°C (in -4H2O)
Erbium(III) nitrate pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 233-436-7

TSCA : Yes

Recommended Storage : Ambient temperatures