missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1130622

 View more versions of this product

Catalog No. AA1130622



Specifications

Specifications

Erbium(III) nitrate pentahydrate
(REO)
MFCD00149691
acmc-20al2v
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
443.346
443.35 (353.27 Anhydrous)
99.99%
Odorless
130°C (in -4H2O)
10031-51-3
ErH10N3O14
UN1477
LWHHUEHWVBVASY-UHFFFAOYSA-N
erbium(3+);trinitrate;pentahydrate
53249207
Crystalline Aggregates
Reacton™
Hygroscopic
100g

Chemical Identifiers

10031-51-3
443.346
LWHHUEHWVBVASY-UHFFFAOYSA-N
53249207
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Er+3]
ErH10N3O14
MFCD00149691
acmc-20al2v
erbium(3+);trinitrate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents